Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad9 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10057077
Description
Rad9 Polyclonal specifically detects Rad9 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
Rad9 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
cell cycle checkpoint control protein RAD9A, DNA repair exonuclease rad9 homolog A, EC 3.1.11.2, hRAD9, RAD9, RAD9 (S. pombe) homolog, RAD9 homolog A (S. pombe) | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Dog, Mouse and Rat. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/mL, Peptide ELISA Detection limit 1:32000 | |
NP_004575.1 | |
RAD9A | |
Peptide with sequence C-QGPSPVLAEDSEGE corresponding to C-Terminus according to NP_004575.1. | |
0.1 mg | |
Apoptosis, Cancer, Checkpoint signaling, DNA Repair, Tumor Suppressors | |
5883 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction