Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177978
Description
Rad9 Polyclonal specifically detects Rad9 in Yeast samples. It is validated for Western Blot, ELISA.Specifications
Rad9 | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
cell cycle checkpoint control protein RAD9A, DNA repair exonuclease rad9 homolog A, EC 3.1.11.2, hRAD9, RAD9, RAD9 (S. pombe) homolog, RAD9 homolog A (S. pombe) | |
Rabbit | |
148.4 kDa | |
0.1 mg | |
Apoptosis, Cancer, Checkpoint signaling, Chromatin Research, DNA Repair, Tumor Suppressors | |
5883 | |
Yeast | |
IgG |
Western Blot, ELISA | |
1 mg/ml | |
Western Blot 1:100-1:500, ELISA 1:5000 | |
Q99638 | |
RAD9A | |
This affinity purified Rad9 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 1120-1145 from the aa1309 yeast Rad9 protein conjugated to KLH. (Uniprot: P14737) | |
Affinity Purified | |
RUO | |
Primary | |
This affinity purified antibody is directed against an internal sequence of yeast Rad9 at the S1129 residue. This pan reactive polyclonal antibody reacts equally with both phosphorylated and non- phosphorylated yeast Rad9 at S1129. No reactivity is expected against human and mouse homologs. Reactivity to Rad9 from others sources is unknown. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction