Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD9 Monoclonal Antibody (OTI7B11)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524923

Catalog No. PIMA524923


Only null left
Add to Cart

Description

Description

RAD9 Monoclonal Antibody for Western Blot, IHC (P)

RAD9 is a component of the 9-1-1 cell cycle checkpoint response complex that plays a major role in DNA repair. The 9-1-1 complex is recruited to DNA lesions upon damage by the RAD17-replication factor C clamp loader complex. RAD9 acts as a sliding clamp platform on DNA for several proteins involved in long-patch base excision repair.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RAD9
Monoclonal
2 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q99638
RAD9A
Full-length protein expressed in 293T cell transfected with human RAD9A expression vector.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI7B11
Unconjugated
RAD9A
Cell cycle checkpoint control protein RAD9A; chromatin-binding protein RAD9; DNA repair exonuclease rad9 homolog A; DNA repair protein RAD9; hRAD9; mRAD9; QtsA-19913; Rad9; RAD9 checkpoint clamp component A; RAD9 homolog A; RAD9 homolog A (S. pombe); Rad9a; Rad9-like protein; rCG47324-like; unnamed protein product; YD9934.02C; YDR217C
Mouse
Affinity Chromatography
RUO
5883
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.