Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad9, p Ser1260 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177981
Description
Rad9 Polyclonal specifically detects Rad9 in Yeast samples. It is validated for Western Blot, ELISA.Specifications
Rad9 (p Ser1260) | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
cell cycle checkpoint control protein RAD9A, DNA repair exonuclease rad9 homolog A, EC 3.1.11.2, hRAD9, RAD9, RAD9 (S. pombe) homolog, RAD9 homolog A (S. pombe) | |
Rabbit | |
148.4 kDa | |
0.1 mg | |
Apoptosis, Cancer, Checkpoint signaling, Chromatin Research, DNA Repair, Phospho Specific, Tumor Suppressors | |
5883 | |
Yeast | |
IgG |
Western Blot, ELISA | |
0.41 mg/ml | |
Western Blot 1:500-1:2000, ELISA 1:5000 | |
NP_004575 | |
RAD9A | |
This affinity purified Rad9 [p Ser1260] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to phosphorylated form of an internal region near aa 1225-1275 from the aa1309 yeast Rad9 protein conjugated to KLH. (Uniprot: P14737) | |
Affinity Purified | |
RUO | |
Primary | |
This affinity purified antibody is directed against the phosphorylated form of yeast Rad9 at the pS1260 residue. The resultant affinity purified antibody is phospho specific to yeast pS1260. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction