Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAD9B Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326072MFV500
Description
RAD9B Polyclonal antibody specifically detects RAD9B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
RAD9B | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
cell cycle checkpoint control protein RAD9B, cell cycle checkpoint control protein RAD9B homolog, DNA repair exonuclease rad9 homolog B, FLJ40346, hRAD9B, MGC75426, RAD9 homolog B (S. pombe) | |
Produced in rabbits immunized with E. coli-derived Human RAD9B fragment | |
0.1 mL | |
Cell Cycle and Replication | |
144715 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction