Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD9B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113981
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113981 Supplier Invitrogen™ Supplier No. PA5113981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATHAPISIYF DFPGKPLALS IDDMLVEANF ILATLADEQS RASSPQSLCL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 93%; Horse: 93%; Human: 100%; Pig: 86%; Rabbit: 79%.

RAD9B gene ontology (GO) annotations include DNA repair; DNA replication; cellular response to ionizing radiation; intra-S DNA damage checkpoint; regulation of signal transduction by p53 class mediator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD9B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RAD9B
Gene Accession No. Q6WBX8
Gene Alias A630082N15Rik; BC021784; Cell cycle checkpoint control protein RAD9B; cell cycle checkpoint control protein RAD9B homolog; DNA repair exonuclease rad9 homolog B; hRAD9B; mRAD9B; RAD9 checkpoint clamp component B; RAD9 homolog B; RAD9B
Gene Symbols RAD9B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Human RAD9B (aa 239-288).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144715
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.