Learn More
Description
This product is supplied as a TFA salt, the stoichiometry of which is batch dependant. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content.
Specifications
Specifications
| Quantity | 5 mg |
| Target | Fluorescent Probes for Imaging Bacteria |
| Molecular Formula | C28H28N4O6 |
| Formula Weight | 3081 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

