Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RadHalo Spectroscopic Area Monitor Radio Communication Options

For use with RadHalo Spectroscopic Area Monitor models with the radio option

Supplier:  Thermo Scientific™ MTXBR24L

 View more versions of this product

Catalog No. 19-175-824


Description

Description

Matrix Base Radio and Matrix Radio Repeater Include rugged IP67 housing, 2.4 GHz radio, external antenna connection, power connection, ethernet connection and wall mount Require an appropriate 2.4 GHz antenna kit Do not include a pole mounting kit or screws/anchors for wall mount The Matrix Base Radio and Matrix Radio Repeater Pole Mount Kit includes complete kit for mounting radios to a pole up to 12 inches in diameter.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Matrix Base Radio - Single Laird 2.4GHz radio in IP67 rated housing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.