Learn More
MilliporeSigma™ Supelco™ radiello™ Cartridge Adsorbents For Sampling Anaesthetic Gases/Vapors, Matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh)
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
20/Pk.
Description
Several different radiello™ cartridge adsorbents are available. Each of which are specific for sampling different classes of compounds. The dimensions for each cartridge are 60 mm L x 4.8 or 5.8 mm D. Each cartridge is designed for one time use with the exception of radiello™ thermal desorption (TD) cartridge adsorbents. Each cartridge arrives in a sealed glass or plastic tube wrapped in a transparent thermally sealed polyethylene (PE) bag. The same sealed tube can be used to store the cartridge after sampling prior to desorption/analysis. A sufficient number of adhesive barcode labels are included with every pack of cartridges for easy tracking of sampling date and time. Note: To conduct air sampling using the radiello™ system, radiello™ diffusive bodies and triangular support plates must be ordered (separately) in addition to cartridge adsorbents. radiello™ Starter Kits (RADxxxS) include all the necessary parts to complete a sampling.
Specifications
Specifications
Pore Size | 30 to 50 Mesh |
Product Line | radiello |
Quantity | 20/Pk. |
radiello is a trademark of Institi Clinici Scientifici Maugeri