Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ radiello™ Passive Diffusive Air Samplers Accessories
SDP

Catalog No. 06611844 Shop All Restek Corporation Products
Change view
Click to view available options
Type:
Caps
Polypropylene containers for ready-to-use sampler
Sterile sampler for anesthetic gases and vapors
Vertical Adaptors for Personal Sampling
Vertical snap ring adaptors for ready-to-use sampler
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-611-844 Sterile sampler for anesthetic gases and vapors
06-611-840 Vertical Adaptors for Personal Sampling
06-611-841 Vertical snap ring adaptors for ready-to-use sampler
06-611-842 Caps
06-611-843 Polypropylene containers for ready-to-use sampler
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 06-611-844 Supplier Restek™ Supplier No. RAD125
Only null left
Add to Cart
Add to Cart

Higher uptake rate and capacity, for any given length of sampling without bias from back diffusion. Versatile & easy to use for a wide range of chemical classes. Water repellant body and reusable holders. No pump required.

  • Versatile radiello™ passive diffusive air samplers—now available from Restek™ —feature a unique 3D radial design that provides several performance advancements compared to standard 2D flat axial air samplers
  • These samplers are a reliable, cost-effective alternative that offers both high sensitivity for low-level detection in short sampling periods (down to 15 min), as well as higher capacity so accurate results can be obtained over longer (30 day) sampling times
  • Because the radial design results in a higher rate of uptake and larger capacity, radiello™ air samplers are a versatile solution that allows labs to sample very efficiently for short durations or sample for longer periods without bias from analyte breakthrough or back diffusion
  • These air samplers are available for a wide range of chemical classes, so the choice of sampling material can be tailored to specific applications
  • Ideal for labs supporting environmental monitoring, industrial hygiene, and any other ambient (indoor/outdoor) air testing, radiello™ air samplers from Restek are recommended when a reliable source of versatile, high-performing, easy-to-use air samplers is required
  • Solvent and thermal desorption options available
  • Features water repellant diffusive body and reusable holders
  • Easy to use—no pump required

Specifications

Color Blue
For Use With (Equipment) radiello Passive Air Samplers
Material Mixed
For Use With (Application) Personal Air Sampling
Quantity 10
Type Sterile sampler for anesthetic gases and vapors
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.