Learn More
MilliporeSigma™ Supelco™ radiello™ Triangular Support Plate
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
20/Pk.
Description
Each support plate is threaded for easy diffusive body attachment. Each plate includes a clip and transparent adhesive pocket to hold the radiello™ barcode label.
Specifications
Specifications
| For Use With (Application) | Air Monitoring |
| Quantity | 20/Pk. |
radiello is a trademark of Institi Clinici Scientifici Maugeri