Learn More
Invitrogen™ RADIL Polyclonal Antibody

Description
RADIL Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | RADIL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | RADIL |
| Gene Accession No. | Q96JH8 |
| Gene Alias | AI536456; D930005D10Rik; KIAA1849; Radil; Rap associating with DIL domain; Rap GTPase interactor; Ras association and DIL domains; ras-associating and dilute domain-containing protein; RASIP2 |
| Gene Symbols | RADIL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.