Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RADIL Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | RADIL |
---|---|
Concentration | 0.43 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2810
![]() ![]() |
Proteintech Group Inc
202841AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Radil is a broadly expressed adapter protein that contains a RA-like domain, a PDZ domain and a DIL domain. Like RAPL and RIAM, Radil interacts preferentially with active GTP-bound Rap1, and overexpression of Radil enhances integrin-mediated adhesion. In addition, Radil knock down inhibits cell-substrate adhesion in HT1080, AD293, and NMuMG cells and results in migration defects in SW1353 and zebrafish neural crest cells. Radil regulates neutrophil adhesion by specifically enhancing beta2-integrin activation, as well as focal adhesion kinase (FAK) and paxillin phosphorylation.Specifications
RADIL | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q96JH8 | |
55698 | |
RADIL Fusion Protein Ag13996 | |
Primary | |
-20°C | |
RADIL |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA1849, RADIL | |
RADIL | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title