Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Radixin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529951
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529951 Supplier Invitrogen™ Supplier No. PA529951
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A549, HeLa, HepG2, HCT116, MDA-MB-231. Predicted reactivity: Mouse (96%), Rat (94%), Pig (97%), Chicken (95%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Radixin is a cytoskeletal protein that may be important in linking actin to the plasma membrane. It is highly similar in sequence to both ezrin and moesin. The radixin gene has been localized by fluorescence in situ hybridization to 11q23. A truncated version representing a pseudogene (RDXP2) was assigned to Xp21.3. Another pseudogene that seemed to lack introns (RDXP1) was mapped to 11p by Southern and PCR analyses. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, May 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Radixin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Rdx
Gene Accession No. P35241
Gene Alias AA516625; DFNB24; ESP10; radixin; radixin isoform; Rdx
Gene Symbols Rdx
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Radixin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315655, 5962
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.