Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Radretumab Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA541936
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541936 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541936 Supplier Invitrogen™ Supplier No. MA541936
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Radretumab is a biosimilar that targets FN EDB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Radretumab
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation 30mM histidine, PBS with 10% sucrose, 0.02% Tween 80 and no preservative; pH 5.8
Gene Alias FN EDB; L19-131 I; L19-SIP
Host Species Human
Immunogen Human FN EDB.
Purification Method Metal-chelate chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype scFv κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.