Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rae-1 Antibody (199215), PE/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1758PECY7
Description
Rae-1 Monoclonal antibody specifically detects Rae-1 in Mouse samples. It is validated for Flow CytometrySpecifications
Rae-1 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8480 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Flow Cytometry | |
199215 | |
Flow Cytometry | |
dJ481F12.3;dJ800J21.1;Gle2;MIG14;Mnrp41;mRNA export factor RAE1;MRNP41;RAE1 | |
BaF3 mouse pro-B cell line transfected with Rae-1 beta and Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta | |
0.5 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction