Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rae-1 epsilon Antibody (205001) [DyLight 350], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1135D
Description
Rae-1 epsilon Monoclonal antibody specifically detects Rae-1 epsilon in Mouse samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOFSpecifications
Rae-1 epsilon | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
BaF3 mouse pro-B cell line transfected with mouse Rae-1 epsilon | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
205001 | |
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction