Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rae-1 Rat anti-Mouse, Brilliant Violet 421, Clone: 186107, BD OptiBuild™

Catalog No. BDB748076 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB748076 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB748076 Supplier BD Biosciences Supplier No. 748076
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Rae-1 Monoclonal antibody specifically detects Rae-1 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rae-1
Applications Flow Cytometry
Classification Monoclonal
Clone 186107
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Host Species Rat
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Rae-1
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.