Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAE1 delta Monoclonal Antibody (186107)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA523889
Description
This antibody recognizes Rae-1 alpha, beta, gamma, gamma and epsilon. Reconstitute at 0.5 mg/mL in sterile PBS.
Rae-1 delta is one of several known murine NKG2D ligands that include RAE-1 molecules, H60 and MULT-1. Expression of NKG2D ligands is low or absent on normal adult tissues. However, stressed or transformed cells express NKG2D ligands which in turn activates NK cells tumoricidal activity through NKG2D. Until now, the expression of NKG2D ligands has been mainly studied with NKG2D tetramers which recognizes all NKG2D ligands.
Specifications
RAE1 delta | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9JI58 | |
Raet1d | |
Mouse myeloma cell line NS0-derived recombinant mouse Rae-1delta Leu29-Ser227. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
186107 | |
Unconjugated | |
Raet1d | |
RAE-1delta; RAE-1-delta; Raet1d; retinoic acid early transcript 1D; retinoic acid early transcript delta; retinoic acid early-inducible protein 1-delta | |
Rat | |
Protein A/G | |
RUO | |
56554 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction