Learn More
RAE1 delta Monoclonal Antibody (RD-41), eBioscience™
Description
RAE1 delta Monoclonal Antibody for IHC (F), Flow, ELISA, FN
Specifications
Specifications
| Antigen | RAE1 delta |
| Applications | ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | RD-41 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Raet1d |
| Gene Accession No. | Q9JI58 |
| Gene Alias | RAE-1delta; RAE-1-delta; Raet1d; retinoic acid early transcript 1D; retinoic acid early transcript delta; retinoic acid early-inducible protein 1-delta |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.