Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAE1 gamma Monoclonal Antibody (CX1), Biotin, eBioscience™
GREENER_CHOICE

Catalog No. 5011977
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-119-77 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-119-77 Supplier Invitrogen™ Supplier No. 13588181
  • $99.75
    $74.65 / Each of 1

    Save $25.10

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

The CX1 monoclonal antibody reacts with the mouse retinoic acid early inducible gamma (RAE-1gamma) and weakly cross-reacts with the RAE-1alpha and beta, but not with delta and ε. These 5 members of the RAE-1 family, along with another protein encoded by the minor histocompatibility gene complex called H60 have been identified as ligands for the mouse NKG2D molecule. These ligands are normally absent from adult tissue, however the presence of retinoic acid or certain disease conditions (such as tumors) can induce up-regulation of this protein at the cell surface. RAE-1alpha, -beta, and -gamma and H60 are expressed in BALB/c mice, but not in C57BL/6, whereas RAE-1delta, and -ε are expressed in C57BL/6 mice. It is reported that in NOD mice, both NKG2D and its ligand RAE-1 are expressed on the same NK cells, resulting in self-modulation of NKG2D expression and function. Applications Reported: The CX1 antibody has been reported for use in flow cytometric analysis. Applications Tested: The CX1 antibody has been tested by flow cytometric analysis of YAC-1 cell line and mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of intere...

Mutations in the Schizosaccharomyces pombe Rae1 and Saccharomyces cerevisiae Gle2 genes have been shown to result in accumulation of poly-containing mRNA in the nucleus, suggesting that the encoded proteins are involved in RNA export. The protein encoded by this gene is a homolog of yeast Rae1. It contains four WD40 motifs, and has been shown to localize to distinct foci in the nucleoplasm, to the nuclear rim, and to meshwork-like structures throughout the cytoplasm. This gene is thought to be involved in nucleocytoplasmic transport, and in directly or indirectly attaching cytoplasmic mRNPs to the cytoskeleton. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAE1 gamma
Applications Flow Cytometry
Classification Monoclonal
Clone CX1
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Raet1c
Gene Accession No. O08604
Gene Alias dJ481F12.3; dJ800J21.1; MIG14; Mnrp41; mRNA export factor; mRNA export protein; MRNP41; RAE-1gamma; RAE-1-gamma; Raet1c; retinoic acid early transcript gamma; retinoic acid early-inducible protein 1-gamma; RP4-800J21.2
Gene Symbols Raet1c
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19370
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.