Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rae1 Mouse anti-Human, Unconjugated, Clone: 14G7.1, MilliporeSigma™

Catalog No. MABC123325 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1233-25 25 μL
MABC1233-100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1233-25 Supplier MilliporeSigma Supplier No. MABC123325UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Rae1, clone 14G7.1, Cat. No. MABC1233, is a mouse monoclonal antibody that detects mRNA export factor (Rae1) and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rae1
Applications Western Blot
Classification Monoclonal
Clone 14G7.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias mRNA export factor;Rae1 protein homolog;mRNA-associated protein mrnp 41
Gene Symbols RAE1;MRNP41
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 179 amino acids from the C-terminal half of human mRNA export factor (Rae1).
Molecular Weight of Antigen ∽40 kDa observed; 40.97 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001015885
Test Specificity Clone 14G7.1 specifically detects human mRNA export factor (Rae1). it targets an epitope with in the C-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.