Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541738 Supplier Invitrogen™ Supplier No. PA541738
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EQLDQPISAC CFNHNGNIFA YASSYDWSKG HEFYNPQKKN YIFLRNAAEE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

Mutations in the Schizosaccharomyces pombe Rae1 and Saccharomyces cerevisiae Gle2 genes have been shown to result in accumulation of poly-containing mRNA in the nucleus, suggesting that the encoded proteins are involved in RNA export. The protein encoded by this gene is a homolog of yeast Rae1. It contains four WD40 motifs, and has been shown to localize to distinct foci in the nucleoplasm, to the nuclear rim, and to meshwork-like structures throughout the cytoplasm. This gene is thought to be involved in nucleocytoplasmic transport, and in directly or indirectly attaching cytoplasmic mRNPs to the cytoskeleton. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RAE1
Gene Accession No. P78406
Gene Alias 3230401I12Rik; 41; D2Ertd342e; dJ481F12.3; dJ800J21.1; FLJ30608; HGNC:9828; homolog of yeast Rae1 (Bharathi) mRNA-associated protein of 41 kDa (Kraemer); MGC117333; MGC126076; MGC126077; MIG14; migration-inducing gene 14; MNRP; Mnrp41; mRNA export factor; mRNA export factor-like protein; mRNA export protein; mRNA-associated protein MRNP 41; mRNA-binding protein, 41-kD; MRNP41; OTTHUMP00000166110; rae1; RAE1 (RNA export 1, S.pombe) homolog; rae1 protein; rae1 protein homolog; RAE1 RNA export 1 homolog; RAE1 RNA export 1 homolog (S. pombe); ribonucleic acid export 1; RP4-800J21.2; zgc:56449; zgc:77723
Gene Symbols RAE1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human RAE1 (aa 312-361).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8480
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.