Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Raf-1, p Ser621 Antibody (SR2183), Novus Biologicals™
SDP

Catalog No. NB286731 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB286731 100 μg
NB286990 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB286731 Supplier Novus Biologicals Supplier No. NBP321572100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Raf-1 Monoclonal antibody specifically detects Raf-1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Raf-1
Applications Western Blot
Classification Monoclonal
Clone SR2183
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias cRaf, c-Raf, CRAFraf proto-oncogene serine/threonine protein kinase, EC 2.7.11, EC 2.7.11.1, NS5, Proto-oncogene c-RAF, RAF, RAF proto-oncogene serine/threonine-protein kinase, Raf-1Oncogene RAF1, v-raf-1 murine leukemia viral oncogene homolog 1
Host Species Rabbit
Immunogen A synthesized peptide derived from human phospho-Raf-1 [Ser621] (Uniprot #: P04049)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, mTOR Pathway, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5894
Target Species Human, Mouse, Rat
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.