Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAF1 Rabbit anti-Human, Unconjugated, Clone: IOF-18, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20562
Description
RAF1 Monoclonal antibody specifically detects RAF1 in Human samples. It is validated for WB-Ce.Specifications
RAF1 | |
Monoclonal | |
Unconjugated | |
RAF1 | |
Rabbit | |
100 μg | |
5894 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
IOF-18 | |
v-raf-1 murine leukemia viral oncogene homolog 1 | |
CRAF, NS5, Raf-1, c-Raf | |
A synthetic phosphopeptide corresponding to residues surrounding S621 of human RAF1 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction