Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAG1 Monoclonal Antibody (1D9)
GREENER_CHOICE

Catalog No. PIMA517163
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517163 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517163 Supplier Invitrogen™ Supplier No. MA517163
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17163 targets RAG1 in indirect ELISA and WB applications and shows reactivity with Human samples. The MA5-17163 immunogen is purified recombinant fragment of human RAG1 (amino acids: 818-868) expressed in E. Coli. MA5-17163 detects RAG1 which has a predicted molecular weight of approximately 119kDa.

The protein encoded by this gene is involved in activation of immunoglobulin V-D-J recombination. The encoded protein is involved in recognition of the DNA substrate, but stable binding and cleavage activity also requires RAG2. Defects in this gene can be the cause of several diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAG1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Rag1
Gene Accession No. P15918
Gene Alias E3 ubiquitin-protein ligase RAG1; Endonuclease RAG1; MGC43321; Rag1; RAG-1; RAG-1 Antibody; recombinase activating gene 1; recombinase activating protein; recombination activating 1; recombination activating gene 1; recombination activating gene 1 Antibody; recombination activating gene-1; recombination activating protein 1; recombination activation protein 1; recombination-activating gene 1; RING finger protein 74; RING-type E3 ubiquitin transferase RAG1; RNF74; RNF74 Antibody; V(D)J recombination-activating protein 1
Gene Symbols Rag1
Host Species Mouse
Immunogen Purified recombinant fragment of human RAG1 (amino acids: 818-868) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5896
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.