Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585711
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585711 Supplier Invitrogen™ Supplier No. PA585711
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (96%), Rat (95%), Xenopus laevis (84%), Pig (90%), Rabbit (96%), Chicken (86%), Rhesus Monkey (98%). Positive Control: Molt-4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is involved in activation of immunoglobulin V-D-J recombination. The encoded protein is involved in recognition of the DNA substrate, but stable binding and cleavage activity also requires RAG2. Defects in this gene can be the cause of several diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAG1
Applications Western Blot
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Rag1
Gene Accession No. P15918
Gene Alias E3 ubiquitin-protein ligase RAG1; Endonuclease RAG1; MGC43321; Rag1; RAG-1; RAG-1 Antibody; recombinase activating gene 1; recombinase activating protein; recombination activating 1; recombination activating gene 1; recombination activating gene 1 Antibody; recombination activating gene-1; recombination activating protein 1; recombination activation protein 1; recombination-activating gene 1; RING finger protein 74; RING-type E3 ubiquitin transferase RAG1; RNF74; RNF74 Antibody; V(D)J recombination-activating protein 1
Gene Symbols Rag1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human RAG1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5896
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.