Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588678
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588678 Supplier Invitrogen™ Supplier No. PA588678
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAASFPPTLG LSSAPDEIQH PHIKFSEWKF KLFRVRSFEK TPEEAQKEKK DSFEGKPSLE QSPAVLDKAD GQKPVPTQPL LKAHPKFSKK FHDNEKARGK AIHQANLRHL CRICGNSFRA DEHNRRYPVH GPVDGKTLGL LRKKEKRATS WPDLIAKVFR IDVKADVDSI HPTEFCHNCW SIMHRKFSSA PCEVYFPRNV TMEWHPHTPS CDICNTARRG LKRKSLQPNL QLSKKLKTVL DQARQARQRK RRAQARISSK DVMKKIANCS; Positive Samples: Mouse thymus; Cellular Location: Nucleus.

The protein encoded by this gene is involved in activation of immunoglobulin V-D-J recombination. The encoded protein is involved in recognition of the DNA substrate, but stable binding and cleavage activity also requires RAG2. Defects in this gene can be the cause of several diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAG1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Rag1
Gene Accession No. P15919
Gene Alias E3 ubiquitin-protein ligase RAG1; Endonuclease RAG1; MGC43321; Rag1; RAG-1; RAG-1 Antibody; recombinase activating gene 1; recombinase activating protein; recombination activating 1; recombination activating gene 1; recombination activating gene 1 Antibody; recombination activating gene-1; recombination activating protein 1; recombination activation protein 1; recombination-activating gene 1; RING finger protein 74; RING-type E3 ubiquitin transferase RAG1; RNF74; RNF74 Antibody; V(D)J recombination-activating protein 1
Gene Symbols Rag1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human RAG1 (NP_0004391).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19373
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.