Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569206
Description
This target displays homology in the following species: Horse: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
The protein encoded by this gene is involved in activation of immunoglobulin V-D-J recombination. The encoded protein is involved in recognition of the DNA substrate, but stable binding and cleavage activity also requires RAG2. Defects in this gene can be the cause of several diseases.
Specifications
RAG1 | |
Polyclonal | |
Unconjugated | |
Rag1 | |
E3 ubiquitin-protein ligase RAG1; Endonuclease RAG1; MGC43321; Rag1; RAG-1; RAG-1 Antibody; recombinase activating gene 1; recombinase activating protein; recombination activating 1; recombination activating gene 1; recombination activating gene 1 Antibody; recombination activating gene-1; recombination activating protein 1; recombination activation protein 1; recombination-activating gene 1; RING finger protein 74; RING-type E3 ubiquitin transferase RAG1; RNF74; RNF74 Antibody; V(D)J recombination-activating protein 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
19373 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P15919 | |
Rag1 | |
synthetic peptide corresponding to a region of mouse RAG1. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction