Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAG2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP243666
Description
Specifically detects RAG2 in Human, Rat samples, and it is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting
RAG2 Polyclonal specifically detects RAG2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RAG2 | |
Polyclonal | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
RAG-2, recombination activating gene 2, V(D)J recombination-activating protein 2 | |
Rabbit | |
59 kDa | |
0.1 mL | |
Adaptive Immunity, Immunology | |
5897 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7) with 0.01% Thimerosal | |
RAG2 | |
Recombinant protein encompassing a sequence within the center region of human RAG2. The exact sequence is proprietary. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction