Learn More
RAGE Mouse anti-Human, Clone: OTI7F4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810801

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Able to phosphorylate several exogenous substrates and to undergo autophosphorylation.Specifications
RAGE | |
Monoclonal | |
Unconjugated | |
AGER | |
RAGE, RAGE-1, RAGE1, STK30 | |
Mouse | |
Affinity Chromatography | |
RUO | |
177 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI7F4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15109 | |
AGER | |
Human recombit protein fragment corresponding to amino acids 205-419 of human RAGE produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.