Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAGE Rabbit anti-Human, Clone: 8G4, Bioss

Rabbit Monoclonal Antibody
Supplier: Bioss BSM52809R

Description
Able to phosphorylate several exogenous substrates and to undergo autophosphorylation.Specifications
RAGE | |
Monoclonal | |
1 μg/mL | |
TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH | |
Q15109 | |
AGER | |
Human RAGE between 1 to 350 amino acids. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot | |
8G4 | |
Unconjugated | |
AGER | |
advanced glycation end product receptor; advanced glycation end-products receptor; Advanced glycosylation end product-specific receptor; advanced glycosylation end product-specific receptor variant 2; advanced glycosylation end product-specific receptor variant 3; advanced glycosylation end product-specific receptor variant 4; advanced glycosylation end product-specific receptor variant 5; Ager; Rage; RAGE isoform NtRAGE-delta; RAGE isoform sRAGE-delta; RAGE/AGER; RAGE-4 ORF3; receptor for advanced glycation end-products variant 20; Receptor for advanced glycosylation end products; SCARJ1; sRAGE | |
Rabbit | |
Protein A | |
RUO | |
177 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction