Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAGE Rabbit anti-Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB9714IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9714IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9714IMI Supplier MilliporeSigma Supplier No. AB9714I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-RAGE is a highly specific rabbit polyclonal antibody that targets Advanced glycosylation end product-specific receptor and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAGE
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols AGER,RAGE
Host Species Rabbit
Immunogen KLH-cojugated linear peptide corresponding to 20 amino acids from the N-terminal region of human RAGE.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001127
Test Specificity This polyclonal antibody detects Receptor for Advanced Glycosylation End Products (RAGE). This antibody targets an epitope corresponding to 21 amino acids in the N-terminal region of human RAGE. Human, mouse, and rat RAGE display 95% homology in this epitope region.
Target Species Rat, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.