Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse RAGE Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB11795N100UG
This item is not returnable.
View return policy
Description
AGER Monoclonal specifically detects AGER in Mouse samples. It is validated for Flow Cytometry.Specifications
RAGE | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_031451 | |
Ager | |
Mouse myeloma cell line NS0-derived recombinant mouse RAGE Gly23-Leu342 Accession # NP_031451 | |
RUO | |
5891 | |
Mouse | |
IgG2a |
Flow Cytometry | |
697023 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1 | |
Rat | |
100 μg | |
Primary | |
Detects mouse RAGE in direct ELISAs and Western blots. In Western blots, approximately 15% cross-reactivity with recombinant canine RAGE and no cross-reactivity with recombinant human RAGE or recombinant rat RAGE is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction