Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAI1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128745H
Description
RAI1 Polyclonal antibody specifically detects RAI1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
RAI1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
KIAA1820DKFZP434A139, MGC12824, retinoic acid induced 1, retinoic acid-induced protein 1, SMCR, Smith-Magenis syndrome chromosome region, SMSDKFZp434A139 | |
The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human retinoic acid induced 1 using the numbering given in entry NP_109590.3 (GeneID 10743). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
10743 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction