Learn More
Invitrogen™ RAI1 Monoclonal Antibody (6H5)

Description
Peptide Sequence: MQSFRERCGF HGKQQNYQQT SQETSRLENY RQPSQAGLSC DRQRLLAKDY YNPQPYPSYE GGAGTPSGTA AAVAADKYHR GSKALPTQQG LQGRPAFPGY G.
Specifications
Specifications
| Antigen | RAI1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 6H5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RAI1 |
| Gene Accession No. | Q7Z5J4 |
| Gene Alias | Gt1; Kiaa1820; RAI1; retinoic acid induced 1; retinoic acid-induced protein 1; SMCR; Smith-Magenis syndrome chromosome region; SMS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.