Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598558
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598558 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598558 Supplier Invitrogen™ Supplier No. PA598558
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAI1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene is located within the Smith-Magenis syndrome region on chromosome 17. It is highly similar to its mouse counterpart and is expressed at high levels mainly in neuronal tissues. The protein encoded by this gene includes a polymorphic polyglutamine tract in the N-terminal domain. Expression of the mouse counterpart in neurons is induced by retinoic acid. This gene is associated with both the severity of the phenotype and the response to medication in schizophrenic patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAI1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RAI1
Gene Accession No. Q7Z5J4
Gene Alias Gt1; Kiaa1820; RAI1; retinoic acid induced 1; retinoic acid-induced protein 1; SMCR; Smith-Magenis syndrome chromosome region; SMS
Gene Symbols RAI1
Host Species Rabbit
Immunogen Recombinant Human Retinoic acid-induced protein 1 protein (617-823AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10743
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.