Learn More
RAI14/NORPEG Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304853A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
RAI14 plays a role in actin regulation at the ectoplasmic specialization, a type of cell junction specific to testis. Important for establishment of sperm polarity and normal spermatid adhesion. May also promote integrity of Sertoli cell tight junctions at the blood-testis barrier.Specifications
RAI14/NORPEG | |
Polyclonal | |
Unconjugated | |
RAI14 | |
ankyrin repeat and coiled-coil structure-containing protein, DKFZp564G013, KIAA1334, NORPEG, novel retinal pigment epithelial, novel retinal pigment epithelial cell protein, RAI13, retinoic acid-induced protein 14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26064, 75646 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9EP71, Q9P0K7 | |
Rai14 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.