Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAI14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590726
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590726 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590726 Supplier Invitrogen™ Supplier No. PA590726
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGKESVFFAE PPFKAEISSI RENKDRLSDS TTGADSLLDI SSEADQQDLL SLLQAKVASL TLHNKELQDK LQAKSPKEAE ADLSFDSYHS TQTDLGPSLG KPGETSPPDS KSSPSVLIHS LGKSTTDNDV RIQQLQEILQ DLQKRLESSE AERKQLQVEL QSRRAELVCL NNTEISENSS DLSQKLKETQ SKYEEAMKEV LSVQKQMKLG LVSPESMDNY SHFHELRVTE EEINVLKQDL QNALEESERN KEKVRELEEK LVEREKGTVI KPPVEEYEEM KSSYCSVIEN MNKEKAFLFE K; Positive Samples: Mouse lung, Rat lung; Cellular Location: Cytoplasm, Nucleus, cell cortex, cytoskeleton.

RAI14 plays a role in actin regulation at the ectoplasmic specialization, a type of cell junction specific to testis. Important for establishment of sperm polarity and normal spermatid adhesion. May also promote integrity of Sertoli cell tight junctions at the blood-testis barrier.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAI14
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RAI14
Gene Accession No. Q5U312, Q9EP71
Gene Alias 1700008J19Rik; 1700020L11Rik; Ankycorbin; ankyrin repeat and coiled-coil structure-containing protein; DKFZp564G013; Kiaa1334; mKIAA1334; NORPEG; NORPEG-like protein; novel retinal pigment epithelial cell protein; p125; RAI13; Rai14; retinoic acid induced 14; retinoic acid-induced protein 14
Gene Symbols RAI14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 300-600 of human RAI14 (NP_0563922).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294804, 75646
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.