Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAI16 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297743FR
Description
RAI16 Polyclonal specifically detects RAI16 in Human samples. It is validated for Western Blot.Specifications
RAI16 | |
Polyclonal | |
Western Blot | |
family with sequence similarity 160, member B2, FLJ11125, FLJ21801, MGC138352, RAI16, retinoic acid induced 16, Retinoic acid-induced protein 16 | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
64760 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
FHIP2B | |
Produced in rabbits immunized with E. coli-derived Human RAI16 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction