Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAI16 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB047295 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047295 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047295 Supplier Novus Biologicals Supplier No. NBP297743H
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAI16 Polyclonal specifically detects RAI16 in Human samples. It is validated for Western Blot.

Specifications

Antigen RAI16
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias family with sequence similarity 160, member B2, FLJ11125, FLJ21801, MGC138352, RAI16, retinoic acid induced 16, Retinoic acid-induced protein 16
Gene Symbols FHIP2B
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RAI16 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64760
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.