Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAI16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116902 Supplier Invitrogen™ Supplier No. PA5116902
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 293T, A-549, SH-SY5Y, Mouse testis, Rat testis Immunogen sequence: GEAGPCLEYL LQHKILETLC TLGKAEYPPG MRQQVFQFFS KVLAQVQHPL LHYLSVHRPV QKLLRLGGTA SGSVTEKEEV QFTTVLCSKI QQDPELLAYI LEGKKIVGRK KACGEPTALP KDTTSHGDKD CSHDGAPARP QLDGESCGAQ ALNSHMPAET EELDGGTTES NLITSLLGLC QSKKSRVALK AQENLLLLVS MASPAAATYL VQSSACCPAI VRHLCQLYRS.

FAM160B2, also named as RAI16, may play a role in early lens development in mammals and in upregulation of genes in CD34+ cells in coronary artery disease patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAI16
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FAM160B2
Gene Accession No. Q80YR2, Q86V87
Gene Alias FAM160B2; family with sequence similarity 160 member B2; family with sequence similarity 160, member B2; FP13191; G430067P06Rik; protein FAM160B2; RAI16; retinoic acid induced 16; retinoic acid-induced protein 16
Gene Symbols FAM160B2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 74-303 of human FAM160B2 (NP_0735865).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239170, 306015, 64760
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.