Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562305
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562305 Supplier Invitrogen™ Supplier No. PA562305
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKVIVSVEDA VPTIFCGKIK GLSGVSTKNF SFKREDSVLQ GYDINSQGEE SMGNAEPLRK PIKNRSIKLK KVNSQEIH Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 94%.

Retinoic acid plays a critical role in development, cellular growth, and differentiation. The specific function of this retinoic acid-induced gene has not yet been determined but it may play a role in development. The chromosomal location of this gene designates it to be a candidate for diseases such as Nance-Horan syndrome, sensorineural deafness, non-specific X-linked mental retardation, oral-facial-digital syndrome, and Fried syndrome. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAI2
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAI2
Gene Accession No. Q9Y5P3
Gene Alias RAI2; retinoic acid induced 2; retinoic acid-induced protein 2
Gene Symbols RAI2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAI2. Recombinant protein control fragment (Product #RP-101283).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10742
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.