Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAIDD/CRADD Antibody (14G8), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242688AF350
Description
RAIDD/CRADD Monoclonal specifically detects RAIDD/CRADD in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
RAIDD/CRADD | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CRADD | |
Recombinant human RAIDD/CRADD (1-199aa) purified from E. coli | |
100 μL | |
Primary | |
Human | |
IgG1a κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
14G8 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
CASP2 and RIPK1 domain containing adaptor with death domain, Caspase and RIP adapter with death domain, caspase and RIP adaptor with death domain, death domain containing protein CRADD, death domain-containing protein CRADD, MGC9163, RAIDDdeath adaptor molecule RAIDD, RIP-associated ICH1/CED3-homologous protein with death domain, RIP-associated protein with a death domain | |
Mouse | |
Protein A purified | |
Apoptosis, Cancer, Cell Biology, Tumor Suppressors | |
8738.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction