Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAIDD/CRADD Rabbit anti-Human, Rat, Clone: 3Z9N8, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | RAIDD/CRADD |
---|---|
Clone | 3Z9N8 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
RAIDD/CRADD Monoclonal antibody specifically detects RAIDD/CRADD in Human, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
RAIDD/CRADD | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Biology, Tumor Suppressors | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
8738 | |
IgG | |
Affinity purified |
3Z9N8 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
CASP2 and RIPK1 domain containing adaptor with death domain, Caspase and RIP adapter with death domain, caspase and RIP adaptor with death domain, death domain containing protein CRADD, death domain-containing protein CRADD, MGC9163, RAIDDdeath adaptor molecule RAIDD, RIP-associated ICH1/CED3-homologous protein with death domain, RIP-associated protein with a death domain | |
A synthetic peptide corresponding to a sequence within amino acids 100-199 of human RAIDD/CRADD (P78560). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title