Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAIDD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519864
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519864 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519864 Supplier Invitrogen™ Supplier No. PA519864
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In Western blot applications, this antibody detects a band at ~22kDa. A suggested positive control is Hela cell lysate. PA5-19864 can be used with blocking peptide PEP-0004.

The protein encoded by this gene is a death domain (CARD/DD)-containing protein and has been shown to induce cell apoptosis. Through its CARD domain, this protein interacts with, and thus recruits, caspase 2/ICH1 to the cell death signal transduction complex that includes tumor necrosis factor receptor 1 (TNFR1A), RIPK1/RIP kinase, and numbers of other CARD domain-containing proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAIDD
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CRADD
Gene Accession No. P78560
Gene Alias CASP2 and RIPK1 domain containing adaptor with death domain; CASP2 and RIPK1 domain-containing adaptor with death domain; caspase and RIP adapter with death domain; caspase and RIP adaptor with death domain; CRADD; death adaptor molecule RAIDD; death domain containing protein CRADD; death domain-containing protein CRADD; MGC9163; MRT34; RAIDD; RIP-associated ICH1/CED3-homologous protein with death domain; RIP-associated protein with a death domain
Gene Symbols CRADD
Host Species Rabbit
Immunogen A peptide corresponding to amino acids 99 to 117 of human RAIDD.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8738
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.