Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAIDD Recombinant Rabbit Monoclonal Antibody (7M7M6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538174
Description
Positive Samples: 293T, K-562, Rat liver, Rat heart Immunogen sequence: DTFLDSLQEF PWVREKLKKA REEAMTDLPA GDRLTGIPSH ILNSSPSDRQ INQLAQRLGP EWEPMVLSLG LSQTDIYRCK A.
The protein encoded by this gene is a death domain (CARD/DD)-containing protein and has been shown to induce cell apoptosis. Through its CARD domain, this protein interacts with, and thus recruits, caspase 2/ICH1 to the cell death signal transduction complex that includes tumor necrosis factor receptor 1 (TNFR1A), RIPK1/RIP kinase, and numbers of other CARD domain-containing proteins.
Specifications
RAIDD | |
Recombinant Monoclonal | |
0.13 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P78560 | |
CRADD | |
A synthetic peptide corresponding to a sequence within amino acids 70-150 of human CRADD (P78560). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
7M7M6 | |
Unconjugated | |
CRADD | |
CASP2 and RIPK1 domain containing adaptor with death domain; CASP2 and RIPK1 domain-containing adaptor with death domain; caspase and RIP adapter with death domain; caspase and RIP adaptor with death domain; CRADD; death adaptor molecule RAIDD; death domain containing protein CRADD; death domain-containing protein CRADD; MGC9163; MRT34; RAIDD; RIP-associated ICH1/CED3-homologous protein with death domain; RIP-associated protein with a death domain | |
Rabbit | |
Affinity chromatography | |
RUO | |
314756, 8738 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction