Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rainbow Fraction™ Tiles
SCI_ED

Catalog No. s04322
Click to view available options
:
Rainbow Fraction Tiles w/ Tray
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.

Visual, hands-on learning tool promotes understanding of fractional parts.

The color-coded plastic tiles enable students to manipulate parts of a whole and grasp the relationship between the pieces. The 51 proportional pieces represent one whole and fractional parts down to one-twelfth; all fit on a plastic tray (included).

Objectives:

  • Build confidence in working with fractions and mixed numbers
  • Recognize equivalent fractions
  • Problem-solve employing addition and subtraction
  • Demonstrate a number sense for fractions

Specifications

Grade Level Elementary, Intermediate - Middle School
Grade 1 to 8
For Use With (Application) Visual, hands-on learning tool promotes understanding of fractional parts
Includes 51 proportional pieces that represent one whole and fractional parts down to one-twelfth; Plastic tray
Description Rainbow Fraction™ Tiles
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.