Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RALA Mouse anti-Human, Monkey, Mouse, Clone:4G8B11, Abnova™

Catalog No. 89333059 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-059 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-059 Supplier Abnova Corporation Supplier No. MAB16742
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human RALA.

The product of this gene belongs to the small GTPase superfamily, Ras family of proteins. GTP-binding proteins mediate the transmembrane signaling initiated by the occupancy of certain cell surface receptors. This gene encodes a low molecular mass ras-like GTP-binding protein that shares about 50% similarity with other ras proteins. [provided by RefSeq]

Specifications

Antigen RALA
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4G8B11
Conjugate Unconjugated
Description v-ral simian leukemia viral oncogene homolog A (ras related)
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene RALA
Gene Alias MGC48949, RAL
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 71-203 of human RALA from E. coli.
Quantity 100 μg
Gene ID (Entrez) 5898
Target Species Human, Monkey, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.