Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RalA/RalB Antibody (AT41E2), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP288840AF750
Description
RalA/RalB Monoclonal antibody specifically detects RalA/RalB in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
RalA/RalB | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
AT41E2 | |
Western Blot, Flow Cytometry, ELISA | |
MGC48949, RAL, Ras family small GTP binding protein RALA, RAS-like protein A, ras-related protein Ral-A, v-ral simian leukemia viral oncogene homolog A (ras related) | |
Recombinant human RalA/RalB (1-203aa) purified from E. coli | |
0.1 mL | |
Apoptosis, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction | |
5898 | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction